missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(trimethylsilyloxy)propane 98.0+%, TCI America™
Specifications
Specifications
| CAS | 17887-80-8 |
| Color | Colorless |
| Boiling Point | 77°C |
| Molecular Formula | C9H24O2Si2 |
| Quantity | 5 g |
| InChI Key | DWHYMKOUICVFHL-UHFFFAOYSA-N |
| SMILES | C[Si](C)(C)OCCCO[Si](C)(C)C |
| IUPAC Name | trimethyl(3-trimethylsilyloxypropoxy)silane |
| Molecular Weight (g/mol) | 220.459 |
| PubChem CID | 140303 |
| Show More |
Safety and Handling
| TSCA | No |