CAS RN 17797-11-4
IUPAC Name:
2-(4-chlorophenyl)propan-2-amine
Synonyms:
2-(4-chlorophenyl)propan-2-amine
Molecular Weight (g/mol):
169.65
Molecular Formula:
C9H12ClN
InChi Key:
JCUUNSQUTQELEZ-UHFFFAOYSA-N
SMILES:
CC(C)(N)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Promotions Available
1-(4-Chlorophenyl)-1-methylethylamine, 97%
1-(4-Chlorophenyl)-1-methylethylamine, 97%
CAS: 17797-11-4 Molecular Formula: C9H12ClN Molecular Weight (g/mol): 169.65 MDL Number: MFCD08669636 InChI Key: JCUUNSQUTQELEZ-UHFFFAOYSA-N Synonym: 2-4-chlorophenyl propan-2-amine,1-4-chlorophenyl-1-methylethylamine,2-4-chlorophenyl prop-2-ylamine,4-chloro-alpha,alpha-dimethylbenzylamine,4-chlorocumylamine,alpha-amino-4-chlorocumene,2-4-chlorophenyl-2-propanamine,2-?4-chlorophenyl?propan-2-amine,benzenemethanamine,4-chloro-,a,,a-dimethyl,benzenemethanamine,4-chloro-a,a-dimethyl PubChem CID: 11030256 IUPAC Name: 2-(4-chlorophenyl)propan-2-amine SMILES: CC(C)(N)C1=CC=C(Cl)C=C1