accessibility menu, dialog, popup

UserName

HOO(S)SNHOO

CAS RN 177966-60-8

IUPAC Name: (2S)-3-(1-benzothiophen-3-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-(1-benzothiophen-3-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 443.52
Molecular Formula: C26H21NO4S
InChi Key: BQIZNDWONIMCGM-QHCPKHFHSA-N
SMILES: OC(=O)[C@H](CC1=CSC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)