accessibility menu, dialog, popup

UserName

CAS RN 177582-74-0

NH2NHOO

CAS RN 177582-74-0

IUPAC Name: benzyl N-[4-(aminomethyl)cyclohexyl]carbamate
Synonyms: benzyl N-[4-(aminomethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 262.35
Molecular Formula: C15H22N2O2
InChi Key: AJYOPGOCTHCBTL-UHFFFAOYSA-N
SMILES: NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results