accessibility menu, dialog, popup

UserName

CAS RN 1768-59-8

SHNNHHN

CAS RN 1768-59-8

IUPAC Name: 1-phenyl-3-(phenylamino)thiourea
Synonyms: 1-phenyl-3-(phenylamino)thiourea
Molecular Weight (g/mol): 243.33
Molecular Formula: C13H13N3S
InChi Key: VGIVYSWGHVFQRP-UHFFFAOYSA-N
SMILES: S=C(NNC1=CC=CC=C1)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results