accessibility menu, dialog, popup

UserName

CAS RN 1760-46-9

OOO

CAS RN 1760-46-9

IUPAC Name: 2,2-diphenylacetyl 2,2-diphenylacetate
Synonyms: diphenylacetic acid, anhydride
Molecular Weight (g/mol): 406.48
Molecular Formula: C28H22O3
InChi Key: YZMRCMTTYLBDPD-UHFFFAOYSA-N
SMILES: O=C(OC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results