accessibility menu, dialog, popup

UserName

CH3CH3NNHH2NS

CAS RN 1752-30-3

IUPAC Name: [(propan-2-ylidene)amino]thiourea
Synonyms: acetone thiosemicarbazone
Molecular Weight (g/mol): 131.2
Molecular Formula: C4H9N3S
InChi Key: FQUDPIIGGVBZEQ-UHFFFAOYSA-N
SMILES: CC(C)=NNC(N)=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results