accessibility menu, dialog, popup

UserName

OSOBrBr

CAS RN 174508-31-7

IUPAC Name: 5,7-dibromo-2H,3H-thieno[3,4-b][1,4]dioxine
Synonyms: 5,7-dibromo-2H,3H-thieno[3,4-b][1,4]dioxine
Molecular Weight (g/mol): 299.96
Molecular Formula: C6H4Br2O2S
InChi Key: FHMRWRBNAIDRAP-UHFFFAOYSA-N
SMILES: BrC1=C2OCCOC2=C(Br)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results