CAS RN 17388-39-5
IUPAC Name:
(4aR,5R,6S)-5-ethenyl-4a-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,4H,4aH,5H,6H-pyrano[3,4-c]pyran-1-one
Synonyms:
swertiamarin swertiamaroside swertiamarine swertiamarin (6CI,7CI,8CI)
Molecular Weight (g/mol):
374.34
Molecular Formula:
C16H22O10
InChi Key:
HEYZWPRKKUGDCR-QBXMEVCASA-N
SMILES:
OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC[C@@]3(O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O
1
–
3
of
3
results
1
phyproof™ Swertiamarine, ≥95% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
3
Swertiamarin 98.0+%, TCI America™
CAS: 17388-39-5 Molecular Formula: C16H22O10 Molecular Weight (g/mol): 374.34 MDL Number: MFCD07783984 InChI Key: HEYZWPRKKUGDCR-QBXMEVCASA-N PubChem CID: 442435 ChEBI: CHEBI:9370 IUPAC Name: (4aR,5R,6S)-5-ethenyl-4a-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,4H,4aH,5H,6H-pyrano[3,4-c]pyran-1-one SMILES: OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC[C@@]3(O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O