accessibility menu, dialog, popup

UserName

OHBrHO

CAS RN 17345-77-6

IUPAC Name: 4-bromobenzene-1,2-diol
Synonyms: 4-bromocatechol C6H5BrO2
Molecular Weight (g/mol): 189.01
Molecular Formula: C6H5BrO2
InChi Key: AQVKHRQMAUJBBP-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results