accessibility menu, dialog, popup

UserName

CAS RN 1732-13-4

IUPAC Name: 1,2,3,6,7,8-hexahydropyrene
Synonyms: 1,2,3,6,7,8-hexahydropyrene
Molecular Weight (g/mol): 208.3
Molecular Formula: C16H16
InChi Key: MBAIEZXRGAOPKH-UHFFFAOYSA-N
SMILES: C1CC2=CC=C3CCCC4=C3C2=C(C1)C=C4
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results