accessibility menu, dialog, popup

UserName

CAS RN 17302-01-1

CH3CH3H3CH3C

CAS RN 17302-01-1

IUPAC Name: 3-ethyl-3-methylheptane
Synonyms: 3-ethyl-3-methylheptane
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: HSOMNBKXPGCNBH-UHFFFAOYSA-N
SMILES: CCCCC(C)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results