CAS RN 17295-26-0
IUPAC Name:
6-(acetyloxy)naphthalene-2-carboxylic acid
Synonyms:
6-(acetyloxy)naphthalene-2-carboxylic acid
Molecular Weight (g/mol):
230.22
Molecular Formula:
C13H10O4
InChi Key:
NFTLBCXRDNIJMI-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=C2C=C(C=CC2=C1)C(O)=O
1
–
1
of
1
results
1
6-Acetoxy-2-naphthoic Acid 98.0+%, TCI America™
CAS: 17295-26-0 Molecular Formula: C13H10O4 Molecular Weight (g/mol): 230.219 MDL Number: MFCD00059556 InChI Key: NFTLBCXRDNIJMI-UHFFFAOYSA-N Synonym: 6-acetoxy-2-naphthoic acid,6-acetyloxy-2-naphthoic acid,2-naphthalenecarboxylic acid, 6-acetyloxy,6-acetyloxy naphthalene-2-carboxylic acid,6-hydroxy-2-naphthoic acid acetate,6-acetoxy-2-naphthoicacid,acmc-209e5j,6-acetoxy-2-naphtoic acid,2-acetoxy-6-naphthoic acid PubChem CID: 87038 IUPAC Name: 6-acetyloxynaphthalene-2-carboxylic acid SMILES: CC(=O)OC1=CC2=C(C=C1)C=C(C=C2)C(=O)O