accessibility menu, dialog, popup

UserName

CAS RN 172669-64-6

ONH2OHONHONH2H2O

CAS RN 172669-64-6

IUPAC Name: 2-(2-aminoacetamido)-4-carbamoylbutanoic acid hydrate
Synonyms: 2-(2-aminoacetamido)-4-carbamoylbutanoic acid hydrate
Molecular Weight (g/mol): 221.21
Molecular Formula: C7H15N3O5
InChi Key: KLFWVRQACSYLOH-UHFFFAOYNA-N
SMILES: O.NCC(=O)NC(CCC(N)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results