accessibility menu, dialog, popup

UserName

CAS RN 17265-14-4

NaNaOOOO

CAS RN 17265-14-4

IUPAC Name: disodium decanedioate
Synonyms: disodium sebacate
Molecular Weight (g/mol): 246.21
Molecular Formula: C10H16Na2O4
InChi Key: NCXUIEDQTCQZRK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CCCCCCCCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results