accessibility menu, dialog, popup

UserName

CAS RN 172612-67-8

CH3SFFFFFFSCH3H3CCH3

CAS RN 172612-67-8

IUPAC Name: 3-[2-(2,4-dimethyl-5-phenylthiophen-3-yl)-3,3,4,4,5,5-hexafluorocyclopent-1-en-1-yl]-2,4-dimethyl-5-phenylthiophene
Synonyms: 3-[2-(2,4-dimethyl-5-phenylthiophen-3-yl)-3,3,4,4,5,5-hexafluorocyclopent-1-en-1-yl]-2,4-dimethyl-5-phenylthiophene
Molecular Weight (g/mol): 548.61
Molecular Formula: C29H22F6S2
InChi Key: DYZAFEDVNIEMEL-UHFFFAOYSA-N
SMILES: CC1=C(C(C)=C(S1)C1=CC=CC=C1)C1=C(C2=C(C)SC(=C2C)C2=CC=CC=C2)C(F)(F)C(F)(F)C1(F)F
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)