accessibility menu, dialog, popup

UserName

CAS RN 17216-62-5

IUPAC Name: 1,3-diethyl 2-(2-cyanoethyl)propanedioate
Synonyms: 1,3-diethyl 2-(2-cyanoethyl)propanedioate
Molecular Weight (g/mol): 213.23
Molecular Formula: C10H15NO4
InChi Key: YJJLOESDBPRZIP-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CCC#N)C(=O)OCC
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)