accessibility menu, dialog, popup

UserName

CAS RN 17200-29-2

IUPAC Name: 5-chloro-1,3-benzoxazole
Synonyms: benzoxazole, 5-chloro-
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: VWMQXAYLHOSRKA-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results