CAS RN 1718-51-0
CAS RN 1718-51-0
IUPAC Name:
4-[(2,3,4,5,6-²H₅)phenyl](²H₉)-1,1'-biphenyl
Synonyms:
4-[(2,3,4,5,6-²H₅)phenyl](²H₉)-1,1'-biphenyl
Molecular Weight (g/mol):
244.4
Molecular Formula:
C18H14
InChi Key:
XJKSTNDFUHDPQJ-WZAAGXFHSA-N
SMILES:
[2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C1=C([2H])C([2H])=C(C([2H])=C1[2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]
1
–
1
of
1
results
1
Fortification Solution, SPEX CertiPrep™
CAS: 1718-51-0 Molecular Formula: C18H14 Molecular Weight (g/mol): 244.395 InChI Key: XJKSTNDFUHDPQJ-WZAAGXFHSA-N PubChem CID: 123128 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene SMILES: C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3