accessibility menu, dialog, popup

UserName

BrBr

CAS RN 171408-84-7

IUPAC Name: 2,7-dibromo-9,9'-spirobi[fluorene]
Synonyms: 2,7-dibromo-9,9'-spirobi[fluorene]
Molecular Weight (g/mol): 474.2
Molecular Formula: C25H14Br2
InChi Key: UPJLZKCEPFAKSH-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results