CAS RN 171408-76-7
IUPAC Name:
7-bromo-9,9'-spirobi[fluorene]
Synonyms:
7-bromo-9,9'-spirobi[fluorene]
Molecular Weight (g/mol):
395.3
Molecular Formula:
C25H15Br
InChi Key:
ONCCVJKFWKAZAE-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
2-Bromo-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
CAS: 171408-76-7 Molecular Formula: C25H15Br Molecular Weight (g/mol): 395.30 MDL Number: MFCD08704217 InChI Key: ONCCVJKFWKAZAE-UHFFFAOYSA-N PubChem CID: 15287522 IUPAC Name: 7-bromo-9,9'-spirobi[fluorene] SMILES: BrC1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12