accessibility menu, dialog, popup

UserName

ClNHN

CAS RN 1707-67-1

IUPAC Name: 2-(2-chlorophenyl)-4,5-diphenyl-1H-imidazole
Synonyms: 2-(2-chlorophenyl)-4,5-diphenyl-1H-imidazole
Molecular Weight (g/mol): 330.82
Molecular Formula: C21H15ClN2
InChi Key: NSWNXQGJAPQOID-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results