accessibility menu, dialog, popup

UserName

FZrFFFFFKK

CAS RN 16923-95-8

IUPAC Name: dipotassium hexafluorozirconiumdiuide
Synonyms: dipotassium hexafluorozirconiumdiuide
Molecular Weight (g/mol): 283.41
Molecular Formula: F6K2Zr
InChi Key: BJZIJOLEWHWTJO-UHFFFAOYSA-H
SMILES: [K+].[K+].F[Zr--](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results