accessibility menu, dialog, popup

UserName

CH3OOOOH3C

CAS RN 1687-29-2

IUPAC Name: 1,2-dimethyl cyclohexane-1,2-dicarboxylate
Synonyms: 1,2-dimethyl cyclohexane-1,2-dicarboxylate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: AIACXWOETVLBIA-UHFFFAOYNA-N
SMILES: COC(=O)C1CCCCC1C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results