accessibility menu, dialog, popup

UserName

NH2HNOCl

CAS RN 1673-47-8

IUPAC Name: 3-chlorobenzohydrazide
Synonyms: benzoylhydrazine, M-chloro
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: PHRDZSRVSVNQRN-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)