accessibility menu, dialog, popup

UserName

CAS RN 1663-45-2

IUPAC Name: [2-(diphenylphosphanyl)ethyl]diphenylphosphane
Synonyms: diphos|dppe|dppe cpd
Molecular Weight (g/mol): 398.43
Molecular Formula: C26H24P2
InChi Key: QFMZQPDHXULLKC-UHFFFAOYSA-N
SMILES: C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)