accessibility menu, dialog, popup

UserName

CAS RN 1655-35-2

IUPAC Name: disodium naphthalene-2,7-disulfonate
Synonyms: disodium naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 332.25
Molecular Formula: C10H6Na2O6S2
InChi Key: XOIWXJSPLXGSLZ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=CC(=CC=C2C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results