accessibility menu, dialog, popup

UserName

CAS RN 1642-54-2

IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid; N,N-diethyl-4-methylpiperazine-1-carboxamide
Synonyms: citric acid; diethylcarbamazine
Molecular Weight (g/mol): 391.42
Molecular Formula: C16H29N3O8
InChi Key: PGNKBEARDDELNB-UHFFFAOYSA-N
SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results