accessibility menu, dialog, popup

UserName

CH3Br

CAS RN 16400-32-1

IUPAC Name: 1-bromopent-2-yne
Synonyms: 1-bromopent-2-yne
Molecular Weight (g/mol): 147.02
Molecular Formula: C5H7Br
InChi Key: VDHGRVFJBGRHMD-UHFFFAOYSA-N
SMILES: CCC#CCBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)