accessibility menu, dialog, popup

UserName

CAS RN 16352-18-4

IUPAC Name: 4,4,5,5-tetramethyl-2-oxo-1,3,2λ⁵-dioxaphospholan-2-ylium
Synonyms: 4,4,5,5-tetramethyl-2-oxo-1,3,2λ⁵-dioxaphospholan-2-ylium
Molecular Weight (g/mol): 163.13
Molecular Formula: C6H12O3P
InChi Key: MWVBQXVOGPKEII-UHFFFAOYSA-N
SMILES: CC1(C)O[P+](=O)OC1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results