accessibility menu, dialog, popup

UserName

CAS RN 163451-82-9

OOHHOOHO

CAS RN 163451-82-9

IUPAC Name: 2-hydroxy-4-(2-hydroxyethoxy)benzoic acid
Synonyms: 2-hydroxy-4-(2-hydroxyethoxy)benzoic acid
Molecular Weight (g/mol): 198.17
Molecular Formula: C9H10O5
InChi Key: INEIMLMPQGJCGK-UHFFFAOYSA-N
SMILES: OCCOC1=CC(O)=C(C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results