CAS RN 163439-82-5
IUPAC Name:
(3R,4R)-1-benzyl-3,4-dihydroxypyrrolidin-1-ium
Synonyms:
(3R,4R)-1-benzyl-3,4-dihydroxypyrrolidin-1-ium
Molecular Weight (g/mol):
194.25
Molecular Formula:
C11H16NO2
InChi Key:
QJRIUWQPJVPYSO-GHMZBOCLSA-O
SMILES:
O[C@@H]1C[NH+](CC2=CC=CC=C2)C[C@H]1O
1
–
1
of
1
results
1
Promotions Available
(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol, 97%
(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol, 97%
CAS: 163439-82-5 MDL Number: MFCD01073894 InChI Key: QJRIUWQPJVPYSO-GHMZBOCLSA-N Synonym: 3r,4r-1-benzylpyrrolidine-3,4-diol,3r,4r---1-benzyl-3,4-pyrrolidindiol,3r,4r---1-benzyl-3,4-pyrrolidinediol,3,4-pyrrolidinediol, 1-phenylmethyl-, 3r,4r,pubchem14982,trans-3,4-dihydroxy-n-benzylpyrrolidine,3r,4r-1-benzyl-3,4-pyrrolidinediol,3r-1-benzylpyrrolidine-3beta,4alpha-diol,3r,4r-rel-1-phenylmethyl-3,4-pyrrolidinediol,3,4-pyrrolidinediol,1-phenylmethyl-, 3r,4r PubChem CID: 7021491 IUPAC Name: (3R,4R)-1-benzylpyrrolidine-3,4-diol SMILES: C1C(C(CN1CC2=CC=CC=C2)O)O