accessibility menu, dialog, popup

UserName

CAS RN 1627844-78-3

H3CONNOCH3CH3(E/Z)

CAS RN 1627844-78-3

IUPAC Name: [4-(decyloxy)-3-methylphenyl][4-(decyloxy)phenyl]diazene
Synonyms: [4-(decyloxy)-3-methylphenyl][4-(decyloxy)phenyl]diazene
Molecular Weight (g/mol): 508.79
Molecular Formula: C33H52N2O2
InChi Key: HBJQHHIKMMWGLL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC1=CC=C(C=C1)N=NC1=CC(C)=C(OCCCCCCCCCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results