CAS RN 162715-14-2
IUPAC Name:
copper(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms:
copper(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol):
623.82
Molecular Formula:
C4CuF12N2O8S4
InChi Key:
UQENANCFJGYGSY-UHFFFAOYSA-N
SMILES:
[Cu++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F