missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Bis(trifluoromethanesulfonyl)imide 96.0+%, TCI America™

Catalog No. C28271G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28271G Supplier TCI America Supplier No. C28271G
Only null left

Specifications

CAS 162715-14-2
Color Blue-Green
Molecular Formula C4H2CuF12N2O9S4
Quantity 1 g
UN Number 1759
Synonym Bis(trifluoromethanesulfonyl)imide Copper(II) Salt, Copper(II) Triflimide
InChI Key SOSOQNLWNNNBFY-UHFFFAOYSA-N
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.O.[Cu+2]
IUPAC Name copper;bis(trifluoromethylsulfonyl)azanide;hydrate
Molecular Weight (g/mol) 641.832
PubChem CID 117065272
Formula Weight 623.82
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Bis(trifluoromethanesulfonyl)imide
Show More Show Less