CAS RN 16239-18-2
IUPAC Name:
1,6-dibromonaphthalen-2-ol
Synonyms:
2-naphthol, 1,6-dibromo-
Molecular Weight (g/mol):
301.97
Molecular Formula:
C10H6Br2O
InChi Key:
VKESFYLPKHQOOA-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C=C(Br)C=CC2=C1Br
1
–
1
of
1
results
1
1,6-Dibromo-2-naphthol 97.0+%, TCI America™
CAS: 16239-18-2 Molecular Formula: C10H6Br2O Molecular Weight (g/mol): 301.97 MDL Number: MFCD00003870 InChI Key: VKESFYLPKHQOOA-UHFFFAOYSA-N Synonym: 1,6-dibromo-2-naphthol,2-naphthalenol, 1,6-dibromo,2-naphthol, 1,6-dibromo,1,6-dibromo-2-hydroxynaphthalene,1,6-dibromo-beta-naphthol,1,6-dibromo-.beta.-naphthol,pubchem22920,2-naphthol,6-dibromo,1,6 dibromo-2-napthol,acmc-1c8jh PubChem CID: 27768 IUPAC Name: 1,6-dibromonaphthalen-2-ol SMILES: OC1=CC=C2C=C(Br)C=CC2=C1Br