CAS RN 161886-96-0
IUPAC Name:
3-(thiophen-3-yl)aniline
Synonyms:
3-(thiophen-3-yl)aniline
Molecular Weight (g/mol):
175.25
Molecular Formula:
C10H9NS
InChi Key:
SHUODWYRFVFGOA-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=C1)C1=CSC=C1
1
–
1
of
1
results
1
3-Thien-3-ylaniline, ≥97%, Thermo Scientific™
CAS: 161886-96-0 Molecular Formula: C10H9NS Molecular Weight (g/mol): 175.25 MDL Number: MFCD06740169 InChI Key: SHUODWYRFVFGOA-UHFFFAOYSA-N Synonym: 3-thien-3-ylaniline,3-thiophen-3-yl aniline,3-3-aminophenyl thiophene,3-thiophen-3-yl-phenylamine,3-3-thienyl aniline,3-3-thienyl phenylamine,3-thiophen-3-ylphenylamine,benzenamine,3-3-thienyl,3-thiophen-3-yl benzenamine,benzenamine, 3-3-thienyl PubChem CID: 4130337 IUPAC Name: 3-thiophen-3-ylaniline SMILES: NC1=CC=CC(=C1)C1=CSC=C1