CAS RN 16165-68-7
IUPAC Name:
diethyl decylphosphonate
Synonyms:
diethyl decylphosphonate
Molecular Weight (g/mol):
278.37
Molecular Formula:
C14H31O3P
InChi Key:
HYQXIVAZQBYBAF-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCP(=O)(OCC)OCC
1
–
1
of
1
results
1
Promotions Available
Diethyl 1-decylphosphonate, 97%
Diethyl 1-decylphosphonate, 97%
CAS: 16165-68-7 Molecular Formula: C14H31O3P Molecular Weight (g/mol): 278.373 MDL Number: MFCD00015593 InChI Key: HYQXIVAZQBYBAF-UHFFFAOYSA-N Synonym: diethyl 1-decylphosphonate,diethyl decylphosphonate,decylphosphonic acid diethyl ester,decyldiethoxyphosphino-1-one,acmc-20al9l,diethyl n-decanephosphonate,diethyl 1-decanephosphonate,diethyl 1-decyl phosphonate,decyl-phosphonic acid diethyl ester,n-decylphosphonic acid diethyl ester PubChem CID: 2773697 IUPAC Name: 1-diethoxyphosphoryldecane SMILES: CCCCCCCCCCP(=O)(OCC)OCC