accessibility menu, dialog, popup

UserName

CAS RN 160976-02-3

FIFBr

CAS RN 160976-02-3

IUPAC Name: 5-bromo-1,3-difluoro-2-iodobenzene
Synonyms: 5-bromo-1,3-difluoro-2-iodobenzene
Molecular Weight (g/mol): 318.89
Molecular Formula: C6H2BrF2I
InChi Key: NLWAKVGODLJALJ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1I
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results