accessibility menu, dialog, popup

UserName

CAS RN 1602-42-2

H2NNH2NHNNOONOOOHO

CAS RN 1602-42-2

IUPAC Name: 5-[(diaminomethylidene)amino]-2-[(2,4-dinitrophenyl)amino]pentanoic acid
Synonyms: 5-[(diaminomethylidene)amino]-2-[(2,4-dinitrophenyl)amino]pentanoic acid
Molecular Weight (g/mol): 340.3
Molecular Formula: C12H16N6O6
InChi Key: GZJXZYUXRVBOAH-UHFFFAOYNA-N
SMILES: NC(N)=NCCCC(NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results