accessibility menu, dialog, popup

UserName

BrBrBrBr

CAS RN 1592-31-0

IUPAC Name: 1,4-bis(dibromomethyl)benzene
Synonyms: 1,4-bis(dibromomethyl)benzene
Molecular Weight (g/mol): 421.75
Molecular Formula: C8H6Br4
InChi Key: VIQBABDKNOOCQD-UHFFFAOYSA-N
SMILES: BrC(Br)C1=CC=C(C=C1)C(Br)Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results