CAS RN 15796-82-4
CAS RN 15796-82-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,4'-Di-tert-butylbenzophenone 95.0+%, TCI America™
CAS: 15796-82-4 Molecular Formula: C21H26O Molecular Weight (g/mol): 294.438 MDL Number: MFCD01319937 InChI Key: YNPFOBWIQVHZMO-UHFFFAOYSA-N Synonym: Bis[4-(tert-butyl)phenyl]methanone PubChem CID: 2758048 IUPAC Name: bis(4-tert-butylphenyl)methanone SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(C)(C)C