accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOH

CAS RN 157634-00-9

IUPAC Name: tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
Synonyms: tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 215.29
Molecular Formula: C11H21NO3
InChi Key: PZTAGFCBNDBBFZ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCCC1CO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)