accessibility menu, dialog, popup

UserName

H3CCH3OO(R)(S)OH(R)OH(R)(R)HOOFFF(Z)(E)

CAS RN 157283-68-6

IUPAC Name: propan-2-yl (5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3R)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]but-1-en-1-yl]cyclopentyl]hept-5-enoate
Synonyms: travoprost|travatan|travatan Z|travoprost (product)|travoprost preparation|travoprost (substance)|alcon brand of travoprost
Molecular Weight (g/mol): 500.56
Molecular Formula: C26H35F3O6
InChi Key: MKPLKVHSHYCHOC-AHTXBMBWSA-N
SMILES: CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)COC1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results