accessibility menu, dialog, popup

UserName

CAS RN 1569-44-4

IUPAC Name: 3-methylhex-5-en-3-ol
Synonyms: 3-methylhex-5-en-3-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: GBARKLJMQRUBKV-UHFFFAOYNA-N
SMILES: CCC(C)(O)CC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)