missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-hexen-3-ol, 98%, Thermo Scientific™

Catalog No. AAB2044706
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2044706 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2044706 Supplier Thermo Scientific Chemicals Supplier No. B2044706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1569-44-4
Density 0.84
Boiling Point 138°C to 139°C
Flash Point 44°C (111°F)
Molecular Formula C7H14O
Quantity 5g
MDL Number MFCD00021826
UN Number UN1987
Synonym 3-methyl-5-hexen-3-ol, methylaethylallylcarbinol, methylethylallylcarbinol, 5-hexen-3-ol, 3-methyl, methylaethylallylcarbinol german, 5-hexen-3-ol, 3-methyl-, 3r, acmc-20m3dr, acmc-1bx1u, 3-methyl-5-hexene-3-ol
InChI Key GBARKLJMQRUBKV-UHFFFAOYNA-N
SMILES CCC(C)(O)CC=C
IUPAC Name 3-methylhex-5-en-3-ol
Molecular Weight (g/mol) 114.19
PubChem CID 15288
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 3-Methyl-5-hexen-3-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECSNumber MP8599000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only