CAS RN 156150-67-3
IUPAC Name:
2-chloro-1-fluoro-4-iodobenzene
Synonyms:
2-chloro-1-fluoro-4-iodobenzene
Molecular Weight (g/mol):
256.44
Molecular Formula:
C6H3ClFI
InChi Key:
OMASDGWBVAVFQZ-UHFFFAOYSA-N
SMILES:
FC1=C(Cl)C=C(I)C=C1
1
–
1
of
1
results
1
Promotions Available
2-Chloro-1-fluoro-4-iodobenzene, 98%
2-Chloro-1-fluoro-4-iodobenzene, 98%
CAS: 156150-67-3 Molecular Formula: C6H3ClFI Molecular Weight (g/mol): 256.443 MDL Number: MFCD00042210 InChI Key: OMASDGWBVAVFQZ-UHFFFAOYSA-N Synonym: 3-chloro-4-fluoroiodobenzene,1-chloro-2-fluoro-5-iodobenzene,2-chloro4-iodofluorobenzene,3-chloro-4-fluoro-1-iodobenzene,1-fluoro-2-chloro-4-iodobenzene,benzene, 2-chloro-1-fluoro-4-iodo,benzene,2-chloro-1-fluoro-4-iodo,2-chloro-1-fluoro-4-iodo-benzene,pubchem3433,acmc-20an6s PubChem CID: 2724605 IUPAC Name: 2-chloro-1-fluoro-4-iodobenzene SMILES: C1=CC(=C(C=C1I)Cl)F