accessibility menu, dialog, popup

UserName

LiH3CCH3OO

CAS RN 15590-62-2

IUPAC Name: lithium(1+) 2-ethylhexanoate
Synonyms: lithium(1+) 2-ethylhexanoate
Molecular Weight (g/mol): 150.15
Molecular Formula: C8H15LiO2
InChi Key: MAZKNBXUMANNDL-UHFFFAOYNA-M
SMILES: [Li+].CCCCC(CC)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results