accessibility menu, dialog, popup

UserName

H3COOH3CNH3COH

CAS RN 15574-49-9

IUPAC Name: ethyl 5-hydroxy-1,2-dimethyl-1H-indole-3-carboxylate
Synonyms: ethyl 5-hydroxy-1,2-dimethylindole-3-carboxylate
Molecular Weight (g/mol): 233.27
Molecular Formula: C13H15NO3
InChi Key: YTBNTDMBGXAOCG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N(C)C2=CC=C(O)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results