accessibility menu, dialog, popup

UserName

ONNNClClClClClCl(E/Z)

CAS RN 154880-05-4

IUPAC Name: 2-[2-(furan-2-yl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Synonyms: 2-[2-(furan-2-yl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 407.88
Molecular Formula: C11H5Cl6N3O
InChi Key: PNDRGJCVJPHPOZ-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C1=NC(=NC(C=CC2=CC=CO2)=N1)C(Cl)(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results